Product Details
- SNP ID
-
rs675
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:116820959 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCTCCAGCTCAGGGAGGGAGAGAG[A/T]CTTGTCCTGGCTCTCTTTCTCCTTG
- Phenotype
-
MIM: 107690
MIM: 107720
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
APOA4
PubMed Links
Gene Details
- Gene
- APOA4
- Gene Name
- apolipoprotein A4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000482.3 |
1203 |
Missense Mutation |
ACT,TCT |
T367S |
NP_000473.2 |
- Gene
- APOC3
- Gene Name
- apolipoprotein C3
There are no transcripts associated with this gene.
View Full Product Details