Product Details

SNP ID
rs180203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:132868089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAGCCCAGTCCACACTCTTCT[C/T]TGATGAACCACTTTTCTTTTCCTAG
Phenotype
MIM: 188450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TG PubMed Links

Gene Details

Gene
TG
Gene Name
thyroglobulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003235.4 Intron NP_003226.4
XM_005251038.4 Intron XP_005251095.1
XM_005251040.4 Intron XP_005251097.1
XM_005251042.4 Intron XP_005251099.1
XM_006716622.3 Intron XP_006716685.1
XM_017013793.1 Intron XP_016869282.1
XM_017013794.1 Intron XP_016869283.1
XM_017013795.1 Intron XP_016869284.1
XM_017013796.1 Intron XP_016869285.1
XM_017013797.1 Intron XP_016869286.1
XM_017013798.1 Intron XP_016869287.1
XM_017013799.1 Intron XP_016869288.1
XM_017013800.1 Intron XP_016869289.1

View Full Product Details