Product Details

SNP ID
rs221834
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:100745552 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCTGTGTCAGAGCCGCTGTATCTCT[C/G]GCCAGTTTGCGTGAATAGGTTATGT
Phenotype
MIM: 602372
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZAN PubMed Links

Gene Details

Gene
ZAN
Gene Name
zonadhesin (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003386.2 Intron NP_003377.2
NM_173059.2 Intron NP_775082.2
XM_011516555.2 Intron XP_011514857.1
XM_017012581.1 Intron XP_016868070.1

View Full Product Details