Product Details
- SNP ID
-
rs199273
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:37452470 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGATCCCCGCGGGCGCCTACAACCT[C/T]GACCCGCAGCGCCCCGTGCACTTCC
- Phenotype
-
MIM: 603963
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ITGA9
PubMed Links
Gene Details
- Gene
- ITGA9
- Gene Name
- integrin subunit alpha 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002207.2 |
149 |
Silent Mutation |
CTC,CTT |
L32L |
NP_002198.2 |
View Full Product Details