Product Details
- SNP ID
-
rs34520922
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:130137245 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CAGGTCTTCAATCCCTGGGAAGACT[A/G]CAAAAGGAGATCCAGAGGAATGCAT
- Phenotype
-
MIM: 612754
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GLRX3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12259552] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLRX3
- Gene Name
- glutaredoxin 3
- Gene
- LOC102724883
- Gene Name
- uncharacterized LOC102724883
There are no transcripts associated with this gene.
View Full Product Details