Product Details

SNP ID
rs2532925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30931386 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCATACCGAATTTTATTTATTGCC[A/G]CTCAGGAGGGTGGGGGCCTGCTGAA
Phenotype
MIM: 613928 MIM: 612802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPCR1 PubMed Links

Gene Details

Gene
DPCR1
Gene Name
diffuse panbronchiolitis critical region 1
There are no transcripts associated with this gene.

Gene
SFTA2
Gene Name
surfactant associated 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205854.2 418 UTR 3 NP_995326.1
Gene
VARS2
Gene Name
valyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

View Full Product Details