Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004686.4 | 3471 | UTR 3 | NP_004677.3 | ||
XM_006716414.3 | 3471 | Intron | XP_006716477.2 | ||
XM_011544692.2 | 3471 | UTR 3 | XP_011542994.1 | ||
XM_011544693.2 | 3471 | Intron | XP_011542995.1 | ||
XM_017013956.1 | 3471 | UTR 3 | XP_016869445.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001145152.1 | 3471 | Intron | NP_001138624.1 | ||
NM_152415.2 | 3471 | Intron | NP_689628.2 | ||
XM_005273400.2 | 3471 | Intron | XP_005273457.1 | ||
XM_005273401.2 | 3471 | Intron | XP_005273458.1 | ||
XM_006716286.1 | 3471 | Intron | XP_006716349.1 | ||
XM_017013021.1 | 3471 | Intron | XP_016868510.1 | ||
XM_017013022.1 | 3471 | Intron | XP_016868511.1 | ||
XM_017013023.1 | 3471 | Intron | XP_016868512.1 | ||
XM_017013024.1 | 3471 | Intron | XP_016868513.1 | ||
XM_017013025.1 | 3471 | Intron | XP_016868514.1 | ||
XM_017013026.1 | 3471 | Intron | XP_016868515.1 |