Product Details

SNP ID
rs35666400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31568821 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATCCTTCGCATATCTCACATGCA[C/T]GTGACACTGAGCCTCAGCCCAGTCT
Phenotype
MIM: 153440 MIM: 601022 MIM: 191160
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100287329 PubMed Links

Gene Details

Gene
LOC100287329
Gene Name
uncharacterized LOC100287329
There are no transcripts associated with this gene.

Gene
LTA
Gene Name
lymphotoxin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000595.3 Intron NP_000586.2
NM_001159740.2 Intron NP_001153212.1
XM_011514615.2 Intron XP_011512917.1
XM_011514616.2 Intron XP_011512918.1
XM_011514617.2 Intron XP_011512919.1
XM_011514618.1 Intron XP_011512920.1
Gene
NFKBIL1
Gene Name
NFKB inhibitor like 1
There are no transcripts associated with this gene.

Gene
TNF
Gene Name
tumor necrosis factor
There are no transcripts associated with this gene.

View Full Product Details