Product Details

SNP ID
rs35778551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26659025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACAGACAGGGAAACCTGAGGAC[A/G]GTGGTTCTCAGACTTTAGGGGCGTT
Phenotype
MIM: 611082
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIAT PubMed Links

Gene Details

Gene
MIAT
Gene Name
myocardial infarction associated transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details