Product Details

SNP ID
rs2236744
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:57561898 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGCAGCCCCTCTCCTACAGACCAG[C/G]TCCTAGTGGAGTAAATGTCCACCTG
Phenotype
MIM: 614168
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
PCK1 PubMed Links

Gene Details

Gene
PCK1
Gene Name
phosphoenolpyruvate carboxykinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002591.3 Intron NP_002582.3
XM_011528839.1 Intron XP_011527141.1

View Full Product Details