Product Details

SNP ID
rs28935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:33013426 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTCACCTTTGGCGCCGAAGCCCC[A/G]TGGGCAGCTTGGGGAAAAGGGGGTT
Phenotype
MIM: 601784
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASIC2 PubMed Links

Gene Details

Gene
ASIC2
Gene Name
acid sensing ion channel subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001094.4 3104 UTR 3 NP_001085.2
NM_183377.1 3104 UTR 3 NP_899233.1

View Full Product Details