Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001284511.1 | Intron | NP_001271440.1 | |||
NM_001284512.1 | Intron | NP_001271441.1 | |||
NM_001284513.1 | Intron | NP_001271442.1 | |||
NM_001324118.1 | Intron | NP_001311047.1 | |||
NM_152341.4 | Intron | NP_689554.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001258450.1 | Intron | NP_001245379.1 | |||
NM_001258451.1 | Intron | NP_001245380.1 | |||
NM_004203.4 | Intron | NP_004194.3 | |||
NM_182687.2 | Intron | NP_872629.1 | |||
XM_011522734.2 | Intron | XP_011521036.1 | |||
XM_011522735.2 | Intron | XP_011521037.1 | |||
XM_011522736.2 | Intron | XP_011521038.1 |