Product Details

SNP ID
rs2237063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:136030447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTCAGGAGTTTGGGGCTCTGGC[A/G]GAGAGGGCCTGCTGGGAGCACATCT
Phenotype
MIM: 601692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TGFBI PubMed Links

Gene Details

Gene
TGFBI
Gene Name
transforming growth factor beta induced
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000358.2 Intron NP_000349.1

View Full Product Details