Product Details
- SNP ID
-
rs35774126
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:39628002 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TTCATTTTTTTTCTTACCTAACCCT[G/T]TAGCCTTATCTCTCTTCTTGGATGA
- Phenotype
-
MIM: 179550
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RALA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74906720] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RALA
- Gene Name
- RALA Ras like proto-oncogene A
View Full Product Details