Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318046.1 | 5742 | Intron | NP_001304975.1 | ||
NM_001318047.1 | 5742 | Intron | NP_001304976.1 | ||
NM_001318048.1 | 5742 | Intron | NP_001304977.1 | ||
NM_007356.2 | 5742 | Intron | NP_031382.2 | ||
XM_011515975.1 | 5742 | UTR 3 | XP_011514277.1 | ||
XM_011515978.1 | 5742 | Intron | XP_011514280.1 | ||
XM_011515979.2 | 5742 | Intron | XP_011514281.1 | ||
XM_011515980.2 | 5742 | Intron | XP_011514282.1 | ||
XM_017011879.1 | 5742 | Intron | XP_016867368.1 | ||
XM_017011880.1 | 5742 | Intron | XP_016867369.1 |