Product Details

SNP ID
rs7220213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30382037 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTCCTATTTTTTTATAGTTGACC[C/G]CTATGCTTCTTTAAGTGAATATACC
Phenotype
MIM: 603102
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPD PubMed Links

Gene Details

Gene
CPD
Gene Name
carboxypeptidase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199775.1 Intron NP_001186704.1
NM_001304.4 Intron NP_001295.2

View Full Product Details