Product Details

SNP ID
rs11728800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:15473372 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTATTACGAATGGTCAGAGAAGAG[C/T]TCTCTGATAATAAAATTTCAGCTGA
Phenotype
MIM: 612013
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CC2D2A PubMed Links

Gene Details

Gene
CC2D2A
Gene Name
coiled-coil and C2 domain containing 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080522.2 Intron NP_001073991.2
NM_001164720.1 Intron NP_001158192.1
NM_020785.2 Intron NP_065836.2
XM_005248177.1 Intron XP_005248234.1
XM_011513872.2 Intron XP_011512174.1
XM_011513874.2 Intron XP_011512176.1
XM_017008482.1 Intron XP_016863971.1

View Full Product Details