Product Details

SNP ID
rs7559066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:48959563 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAAACTAATGCATCTGTATGTGA[C/T]AAGATAAAAATTTTTGATCAACTTG
Phenotype
MIM: 136435
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FSHR PubMed Links

Gene Details

Gene
FSHR
Gene Name
follicle stimulating hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000145.3 5051 Intron NP_000136.2
NM_181446.2 5051 Intron NP_852111.2
XM_011532733.2 5051 UTR 3 XP_011531035.1
XM_011532734.2 5051 UTR 3 XP_011531036.1
XM_011532735.2 5051 UTR 3 XP_011531037.1
XM_011532736.2 5051 UTR 3 XP_011531038.1
XM_011532740.1 5051 Intron XP_011531042.1

View Full Product Details