Product Details
- SNP ID
-
rs848291
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:58161561 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTTGATGGAAAGGTTGTCCACACTG[A/G]GAATTATCACACACTTGATCAGGAA
- Phenotype
-
MIM: 608111
MIM: 602169
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FANCL
PubMed Links
Gene Details
- Gene
- FANCL
- Gene Name
- Fanconi anemia complementation group L
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001114636.1 |
1093 |
Silent Mutation |
TCC,TCT |
S332S |
NP_001108108.1 |
NM_018062.3 |
1093 |
Silent Mutation |
TCC,TCT |
S327S |
NP_060532.2 |
XM_005264395.3 |
1093 |
Silent Mutation |
TCC,TCT |
S327S |
XP_005264452.1 |
XM_005264397.4 |
1093 |
Silent Mutation |
TCC,TCT |
S210S |
XP_005264454.1 |
XM_011532939.2 |
1093 |
Silent Mutation |
TCC,TCT |
S347S |
XP_011531241.1 |
XM_011532940.2 |
1093 |
Silent Mutation |
TCC,TCT |
S342S |
XP_011531242.1 |
XM_011532941.1 |
1093 |
Silent Mutation |
TCC,TCT |
S347S |
XP_011531243.1 |
XM_011532942.2 |
1093 |
Silent Mutation |
TCC,TCT |
S332S |
XP_011531244.1 |
XM_011532944.2 |
1093 |
Silent Mutation |
TCC,TCT |
S230S |
XP_011531246.1 |
XM_011532945.2 |
1093 |
Silent Mutation |
TCC,TCT |
S215S |
XP_011531247.1 |
XM_017004414.1 |
1093 |
Silent Mutation |
TCC,TCT |
S342S |
XP_016859903.1 |
XM_017004415.1 |
1093 |
Intron |
|
|
XP_016859904.1 |
XM_017004416.1 |
1093 |
Silent Mutation |
TCC,TCT |
S210S |
XP_016859905.1 |
- Gene
- VRK2
- Gene Name
- vaccinia related kinase 2
View Full Product Details