Product Details

SNP ID
rs34071316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41927801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTTCCAGGCCCCTTTTACACCT[A/C]TGGGGAGAGCCATCCTATTGCATTT
Phenotype
MIM: 603871 MIM: 123834 MIM: 612915
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BYSL PubMed Links

Gene Details

Gene
BYSL
Gene Name
bystin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004053.3 Intron NP_004044.3
Gene
CCND3
Gene Name
cyclin D3
There are no transcripts associated with this gene.

Gene
MED20
Gene Name
mediator complex subunit 20
There are no transcripts associated with this gene.

View Full Product Details