Product Details

SNP ID
rs11828658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66685507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGGCAGAAGGCGAGTGCCCTC[A/G]CATGGCAGGAGAATGACCCTGAGGC
Phenotype
MIM: 604985
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM4B PubMed Links

Gene Details

Gene
RBM4B
Gene Name
RNA binding motif protein 4B
There are no transcripts associated with this gene.

Gene
SPTBN2
Gene Name
spectrin beta, non-erythrocytic 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006946.2 8363 UTR 3 NP_008877.1
XM_005274192.4 8363 UTR 3 XP_005274249.1
XM_005274193.3 8363 UTR 3 XP_005274250.1
XM_006718669.3 8363 UTR 3 XP_006718732.1
XM_006718671.3 8363 UTR 3 XP_006718734.1
XM_011545216.2 8363 Intron XP_011543518.1
XM_011545217.2 8363 Intron XP_011543519.1
XM_017018174.1 8363 UTR 3 XP_016873663.1
XM_017018175.1 8363 UTR 3 XP_016873664.1
XM_017018176.1 8363 UTR 3 XP_016873665.1
XM_017018177.1 8363 UTR 3 XP_016873666.1
XM_017018178.1 8363 UTR 3 XP_016873667.1

View Full Product Details