Product Details

SNP ID
rs732337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:20249379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGAAGCAGAGGGGAGCCGCTGT[A/T]CTTGCTGTGGCCACCTTCCCTGGAG
Phenotype
MIM: 606551
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LZTS1 PubMed Links

Gene Details

Gene
LZTS1
Gene Name
leucine zipper tumor suppressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021020.3 2510 UTR 3 NP_066300.1
XM_005273394.3 2510 UTR 3 XP_005273451.1
XM_011544383.2 2510 UTR 3 XP_011542685.1
XM_011544384.2 2510 UTR 3 XP_011542686.1
XM_011544385.2 2510 UTR 3 XP_011542687.1
XM_011544386.2 2510 UTR 3 XP_011542688.1

View Full Product Details