Product Details

SNP ID
rs11128570
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:11721527 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGGATAAATATTCACAGATTAAAGC[C/T]CGGAAGGTTCCCAGTCTTCCCAAAG
Phenotype
MIM: 614948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAMM41 PubMed Links

Gene Details

Gene
TAMM41
Gene Name
TAM41 mitochondrial translocator assembly and maintenance homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284401.1 Intron NP_001271330.1
NM_001321294.1 Intron NP_001308223.1
NM_001321295.1 Intron NP_001308224.1
NM_138807.3 Intron NP_620162.1
XM_005264873.3 Intron XP_005264930.1
XM_005264875.3 Intron XP_005264932.1
XM_017005725.1 Intron XP_016861214.1
XM_017005726.1 Intron XP_016861215.1
XM_017005727.1 Intron XP_016861216.1
XM_017005728.1 Intron XP_016861217.1
Gene
VGLL4
Gene Name
vestigial like family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128219.2 Intron NP_001121691.1
NM_001128220.2 Intron NP_001121692.1
NM_001128221.2 Intron NP_001121693.1
NM_001284390.1 Intron NP_001271319.1
NM_001284391.1 Intron NP_001271320.1
NM_014667.3 Intron NP_055482.2
XM_011534267.2 Intron XP_011532569.1
XM_011534268.1 Intron XP_011532570.1
XM_011534269.1 Intron XP_011532571.1
XM_017007541.1 Intron XP_016863030.1
XM_017007542.1 Intron XP_016863031.1

View Full Product Details