Product Details

SNP ID
rs706115
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:33253607 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTACCTCTACTACCTTGTCTGAGGT[C/G]TTAACCTGCCCAGTCCTGAATTCCT
Phenotype
MIM: 601497 MIM: 613929
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BAG1 PubMed Links

Gene Details

Gene
BAG1
Gene Name
BCL2 associated athanogene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172415.1 2737 UTR 3 NP_001165886.1
NM_004323.5 2737 UTR 3 NP_004314.5
Gene
SPINK4
Gene Name
serine peptidase inhibitor, Kazal type 4
There are no transcripts associated with this gene.

View Full Product Details