Product Details

SNP ID
rs6122476
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63557677 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGACCAGCAGGACAGTCAAGGCTG[A/G]TGGCCTCAGGGACTTTGGACTTGGC
Phenotype
MIM: 611265
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FNDC11 PubMed Links

Gene Details

Gene
FNDC11
Gene Name
fibronectin type III domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319152.1 Intron NP_001306081.1
NM_001319153.1 Intron NP_001306082.1
NM_024059.3 Intron NP_076964.1
XM_011529041.2 Intron XP_011527343.1
XM_017028060.1 Intron XP_016883549.1
Gene
HELZ2
Gene Name
helicase with zinc finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037335.2 Intron NP_001032412.2
NM_033405.3 Intron NP_208384.3

View Full Product Details