Product Details
- SNP ID
-
rs7048663
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21461976 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- TGTTTAACAATATAAAGAGTTAAAT[C/G]ATTAAAAATAATGAATACTAAAATT
- Phenotype
-
MIM: 616356
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR31HG
PubMed Links
Gene Details
- Gene
- MIR31HG
- Gene Name
- MIR31 host gene
There are no transcripts associated with this gene.
View Full Product Details