Product Details

SNP ID
rs2465584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:46768192 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCTACCAGAAAGTTCTAGCCCA[G/T]TTTTGATTTTTGGTTCTTGTACATT
Phenotype
MIM: 608065
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC38A4 PubMed Links

Gene Details

Gene
SLC38A4
Gene Name
solute carrier family 38 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143824.1 Intron NP_001137296.1
NM_018018.4 Intron NP_060488.2
XM_005268997.2 Intron XP_005269054.1

View Full Product Details