Product Details
- SNP ID
-
rs4964725
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:108562106 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAATACAAAAGGGAAGATTCAGACT[C/T]AGCGAGCCGGTACCATTCGTCTAGG
- Phenotype
-
MIM: 611911
MIM: 611684
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ISCU
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11113991,rs75704570] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ISCU
- Gene Name
- iron-sulfur cluster assembly enzyme
- Gene
- SART3
- Gene Name
- squamous cell carcinoma antigen recognized by T-cells 3
View Full Product Details