Product Details

SNP ID
rs2301254
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:32436129 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTGGGTTCCAGAGGTCGGGACTGAA[A/G]GCGAACGTCCCTCAGTTGGGTGGCC
Phenotype
MIM: 607102 MIM: 607899
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WT1 PubMed Links

Gene Details

Gene
WT1
Gene Name
Wilms tumor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000378.4 Intron NP_000369.3
NM_001198551.1 Intron NP_001185480.1
NM_001198552.1 Intron NP_001185481.1
NM_024424.3 Intron NP_077742.2
NM_024426.4 Intron NP_077744.3
Gene
WT1-AS
Gene Name
WT1 antisense RNA
There are no transcripts associated with this gene.

View Full Product Details