Product Details

SNP ID
rs1799937
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:32389228 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGACACTTGCAACAAAGAGACAGGC[A/G]CAAGTTCAACTATCAAGGCCCGAGT
Phenotype
MIM: 607102
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WT1 PubMed Links

Gene Details

Gene
WT1
Gene Name
Wilms tumor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000378.4 Intron NP_000369.3
NM_001198551.1 Intron NP_001185480.1
NM_001198552.1 Intron NP_001185481.1
NM_024424.3 Intron NP_077742.2
NM_024426.4 Intron NP_077744.3

View Full Product Details