Product Details

SNP ID
rs3027471
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:49033821 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGTGATCAGGGCCTCTGAGCACAG[T/G]AAGGCAAGAAACACATGCAGATACT
Phenotype
MIM: 314310
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
TFE3 PubMed Links

Gene Details

Gene
TFE3
Gene Name
transcription factor binding to IGHM enhancer 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282142.1 Intron NP_001269071.1
NM_006521.5 Intron NP_006512.2

View Full Product Details