Product Details

SNP ID
rs2208430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94463371 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAACTGAGTAAACTAAACTGAGTG[C/T]TTTTGCCCTGGAATGGTGATGTCTA
Phenotype
MIM: 615677
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINA9 PubMed Links

Gene Details

Gene
SERPINA9
Gene Name
serpin family A member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042518.1 Intron NP_001035983.1
NM_001284275.1 Intron NP_001271204.1
NM_001284276.1 Intron NP_001271205.1
NM_175739.3 Intron NP_783866.2
XM_011536714.2 Intron XP_011535016.1
XM_011536715.2 Intron XP_011535017.1
XM_011536716.2 Intron XP_011535018.1
XM_011536717.1 Intron XP_011535019.1

View Full Product Details