Product Details

SNP ID
rs2269127
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:42657127 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGACGATCGGCTTTGCAGGCTTTGT[A/G]GGCTTTGTGGGGAAGGAGCCCTTTG
Phenotype
MIM: 602973
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928255 PubMed Links

Gene Details

Gene
LOC101928255
Gene Name
uncharacterized LOC101928255
There are no transcripts associated with this gene.

Gene
PDE9A
Gene Name
phosphodiesterase 9A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001567.1 Intron NP_001001567.1
NM_001001568.1 Intron NP_001001568.1
NM_001001569.1 Intron NP_001001569.1
NM_001001570.1 Intron NP_001001570.1
NM_001001571.1 Intron NP_001001571.1
NM_001001572.1 Intron NP_001001572.1
NM_001001573.1 Intron NP_001001573.1
NM_001001574.1 Intron NP_001001574.1
NM_001001575.1 Intron NP_001001575.1
NM_001001576.1 Intron NP_001001576.1
NM_001001577.1 Intron NP_001001577.1
NM_001001578.1 Intron NP_001001578.1
NM_001001579.1 Intron NP_001001579.1
NM_001001580.1 Intron NP_001001580.1
NM_001001581.1 Intron NP_001001581.1
NM_001001582.1 Intron NP_001001582.1
NM_001001583.1 Intron NP_001001583.1
NM_001001584.2 Intron NP_001001584.1
NM_001001585.1 Intron NP_001001585.1
NM_001315533.1 Intron NP_001302462.1
NM_002606.2 Intron NP_002597.1
XM_011529598.2 Intron XP_011527900.1
XM_011529600.2 Intron XP_011527902.1
XM_017028366.1 Intron XP_016883855.1
XM_017028367.1 Intron XP_016883856.1

View Full Product Details