Product Details
- SNP ID
-
rs11626454
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:24081419 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCGCCGCTGCTGCAGCCGCTTGGA[G/T]CGACAGGCCTGCGCGTAGCCGCGGT
- Phenotype
-
MIM: 605688
MIM: 162080
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CPNE6
PubMed Links
Gene Details
- Gene
- CPNE6
- Gene Name
- copine 6
There are no transcripts associated with this gene.
- Gene
- NRL
- Gene Name
- neural retina leucine zipper
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006177.3 |
446 |
Silent Mutation |
CGA,CGC |
R177R |
NP_006168.1 |
XM_005267708.4 |
446 |
Silent Mutation |
CGA,CGC |
R177R |
XP_005267765.1 |
XM_005267709.3 |
446 |
Silent Mutation |
CGA,CGC |
R177R |
XP_005267766.1 |
XM_005267710.3 |
446 |
Silent Mutation |
CGA,CGC |
R177R |
XP_005267767.1 |
XM_011536801.2 |
446 |
Silent Mutation |
CGA,CGC |
R279R |
XP_011535103.2 |
XM_011536802.1 |
446 |
Silent Mutation |
CGA,CGC |
R177R |
XP_011535104.1 |
XM_011536804.2 |
446 |
Silent Mutation |
CGA,CGC |
R177R |
XP_011535106.1 |
XM_011536805.2 |
446 |
Silent Mutation |
CGA,CGC |
R177R |
XP_011535107.1 |
XM_011536806.2 |
446 |
Silent Mutation |
CGA,CGC |
R174R |
XP_011535108.2 |
XM_017021350.1 |
446 |
Silent Mutation |
CGA,CGC |
R72R |
XP_016876839.1 |
View Full Product Details