Product Details

SNP ID
rs71387164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57377866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTTTTTATACTCATCCTCCTT[A/G]AGCTCAAGCTAAGCAGAAGAAAATT
Phenotype
MIM: 601880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CX3CL1 PubMed Links

Gene Details

Gene
CX3CL1
Gene Name
C-X3-C motif chemokine ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304392.1 Intron NP_001291321.1
NM_002996.4 Intron NP_002987.1

View Full Product Details