Product Details
- SNP ID
-
rs907034
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:83954393 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTTTGCCTTTGTGAAATTTGAAA[C/T]ATCAATTTTTAATGTTAACTTCTTT
- Phenotype
-
MIM: 607975
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC105371371
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77523399] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC105371371
- Gene Name
- uncharacterized LOC105371371
There are no transcripts associated with this gene.
- Gene
- OSGIN1
- Gene Name
- oxidative stress induced growth inhibitor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182981.2 |
|
Intron |
|
|
NP_892026.1 |
View Full Product Details