Product Details

SNP ID
rs34563658
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:32106173 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTACATACCTTTTCAGAATTGTTC[A/G]TATTTTTATTTGGATTGTTTGGGTT
Phenotype
MIM: 616319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF138 PubMed Links

Gene Details

Gene
RNF138
Gene Name
ring finger protein 138
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191324.1 Intron NP_001178253.2
NM_016271.4 Intron NP_057355.2
NM_198128.2 Intron NP_937761.1
XM_005258285.1 Intron XP_005258342.1
XM_005258286.1 Intron XP_005258343.1

View Full Product Details