Product Details

SNP ID
rs11679343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86108085 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGTTCAGTGAGCTATAATTGCAC[C/T]ACTTCACTAAAGGCTAGGCAATGGA
Phenotype
MIM: 616404 MIM: 614918
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR1A PubMed Links

Gene Details

Gene
POLR1A
Gene Name
RNA polymerase I subunit A
There are no transcripts associated with this gene.

Gene
PTCD3
Gene Name
pentatricopeptide repeat domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017952.5 Intron NP_060422.4

View Full Product Details