Product Details

SNP ID
rs2367201
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86107076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAGTTTGTGAGGTATAAGTGGG[A/G]TATGTAATTTACAATATGTTTTTAT
Phenotype
MIM: 616404 MIM: 614918
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
POLR1A PubMed Links

Gene Details

Gene
POLR1A
Gene Name
RNA polymerase I subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015425.4 Intron NP_056240.2
Gene
PTCD3
Gene Name
pentatricopeptide repeat domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017952.5 Intron NP_060422.4

View Full Product Details