Product Details

SNP ID
rs2929046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:55453758 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTGGGTACAGGTTCTTGTTCTTG[C/T]AGAGCTGCTCTACCAGTTCTTGGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SBF1P1 PubMed Links

Gene Details

Gene
SBF1P1
Gene Name
SET binding factor 1 pseudogene 1
There are no transcripts associated with this gene.

Gene
XKR4
Gene Name
XK related 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052898.1 Intron NP_443130.1

View Full Product Details