Product Details

SNP ID
rs28372855
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33081183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCGTAGCGAGGGGGCGGGGCCC[C/G]GAAGAGCAGGATGGCGGCGCGGGAC
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
GPATCH1 PubMed Links

Gene Details

Gene
GPATCH1
Gene Name
G-patch domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018025.2 19 UTR 5 NP_060495.2
XM_006723255.3 19 UTR 5 XP_006723318.1

View Full Product Details