Product Details

SNP ID
rs11668576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51334106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGATGACCTATGACGGCACTGAT[G/T]GCCTCAGCCAGGTCTTCCATAGGCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IGLON5 PubMed Links

Gene Details

Gene
IGLON5
Gene Name
IgLON family member 5
There are no transcripts associated with this gene.

Gene
VSIG10L
Gene Name
V-set and immunoglobulin domain containing 10 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163922.1 Intron NP_001157394.1

View Full Product Details