Product Details

SNP ID
rs12642770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154539307 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAAGCATTCTTCCAAAACACATA[C/T]GTTCTAAATGAAAGTCATATCACAC
Phenotype
MIM: 605961
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLRG1 PubMed Links

Gene Details

Gene
PLRG1
Gene Name
pleiotropic regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201564.1 Intron NP_001188493.1
NM_002669.3 Intron NP_002660.1

View Full Product Details