Product Details

SNP ID
rs12603498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49846726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGGAGAGTAAGGATTAGACCC[A/C]GCTGCAGGATGAGGTGCTTTTGGGA
Phenotype
MIM: 607833
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FLJ45513 PubMed Links

Gene Details

Gene
FLJ45513
Gene Name
uncharacterized LOC729220
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242791.1 Intron NP_001229720.1
Gene
TAC4
Gene Name
tachykinin 4 (hemokinin)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077503.1 Intron NP_001070971.1
NM_001077504.1 Intron NP_001070972.1
NM_001077505.1 Intron NP_001070973.1
NM_001077506.1 Intron NP_001070974.1
NM_170685.2 Intron NP_733786.2

View Full Product Details