Product Details

SNP ID
rs822396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186849088 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGAGAAAGAGATCTTTATTTTT[A/G]ACAAAGGGGGAGGAGCCAGAAAACT
Phenotype
MIM: 605441
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADIPOQ PubMed Links

Gene Details

Gene
ADIPOQ
Gene Name
adiponectin, C1Q and collagen domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177800.1 Intron NP_001171271.1
NM_004797.3 Intron NP_004788.1
Gene
ADIPOQ-AS1
Gene Name
ADIPOQ antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details