Product Details

SNP ID
rs595176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:107526686 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAATTTATAATGAAATTTTACAT[A/G]GCATTGAAGGTCTAGTAAATTTAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BBX PubMed Links

Gene Details

Gene
BBX
Gene Name
BBX, HMG-box containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142568.2 Intron NP_001136040.1
NM_001276286.1 Intron NP_001263215.1
NM_020235.6 Intron NP_064620.2
XM_005247642.3 Intron XP_005247699.1
XM_005247643.3 Intron XP_005247700.1
XM_005247644.3 Intron XP_005247701.1
XM_011512999.2 Intron XP_011511301.1
XM_011513000.1 Intron XP_011511302.1
XM_011513001.1 Intron XP_011511303.1
XM_011513002.2 Intron XP_011511304.1
XM_011513003.2 Intron XP_011511305.1
XM_011513004.2 Intron XP_011511306.1
XM_011513005.2 Intron XP_011511307.1
XM_011513006.2 Intron XP_011511308.1
XM_011513007.2 Intron XP_011511309.1
XM_011513010.2 Intron XP_011511312.1
XM_017006881.1 Intron XP_016862370.1
XM_017006882.1 Intron XP_016862371.1
XM_017006883.1 Intron XP_016862372.1

View Full Product Details