Product Details

SNP ID
rs4703800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:80125915 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGAGAAATTAATTCTATTGCCA[C/T]CTATTCATGAGAATCACAAGGCTGA
Phenotype
MIM: 614551
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERINC5 PubMed Links

Gene Details

Gene
SERINC5
Gene Name
serine incorporator 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174071.2 Intron NP_001167542.1
NM_001174072.2 Intron NP_001167543.1
NM_178276.6 Intron NP_840060.1
XM_011543304.1 Intron XP_011541606.1
XM_017009323.1 Intron XP_016864812.1
XM_017009324.1 Intron XP_016864813.1
XM_017009325.1 Intron XP_016864814.1
XM_017009326.1 Intron XP_016864815.1

View Full Product Details