Product Details
- SNP ID
-
rs26077
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:179913137 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGAGTTCCCACAGGAGAATGCACG[C/T]GCGCTGGGCAAGGTGCCACCCTCGG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RNF130
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113779029] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RNF130
- Gene Name
- ring finger protein 130
- Gene
- TBC1D9B
- Gene Name
- TBC1 domain family member 9B
There are no transcripts associated with this gene.
View Full Product Details