Product Details

SNP ID
rs1966574
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:52864139 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAGCAAAGAGAACTCTATAGCCAC[A/G]TCTCTGAAAGTCAAGTATCCCTAAA
Phenotype
MIM: 606427 MIM: 616841
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF320 PubMed Links

Gene Details

Gene
ZNF320
Gene Name
zinc finger protein 320
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207333.2 Intron NP_997216.2
XM_006723059.3 Intron XP_006723122.1
XM_017026393.1 Intron XP_016881882.1
XM_017026394.1 Intron XP_016881883.1
XM_017026395.1 Intron XP_016881884.1
XM_017026396.1 Intron XP_016881885.1
XM_017026397.1 Intron XP_016881886.1
XM_017026398.1 Intron XP_016881887.1
XM_017026399.1 Intron XP_016881888.1
XM_017026400.1 Intron XP_016881889.1
Gene
ZNF468
Gene Name
zinc finger protein 468
There are no transcripts associated with this gene.

View Full Product Details