Product Details

SNP ID
rs4803015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52870658 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCACACCTCTAGTCCCAGCTACT[C/T]GAGGTGCTCAGGCAAAAGAATCACT
Phenotype
MIM: 606427
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF320 PubMed Links

Gene Details

Gene
ZNF320
Gene Name
zinc finger protein 320
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207333.2 Intron NP_997216.2
XM_006723059.3 Intron XP_006723122.1
XM_017026393.1 Intron XP_016881882.1
XM_017026394.1 Intron XP_016881883.1
XM_017026395.1 Intron XP_016881884.1
XM_017026396.1 Intron XP_016881885.1
XM_017026397.1 Intron XP_016881886.1
XM_017026398.1 Intron XP_016881887.1
XM_017026399.1 Intron XP_016881888.1
XM_017026400.1 Intron XP_016881889.1

View Full Product Details